Learn More
Electron Microscopy Sciences Transparent Wafer Container Single, 4", Polypropylene Material, 110 x 11 mm
SDP
Catalog No. 50365013
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Boxes are transparent and made from Anti-Electrostatic Polycarbonate.
Use: 4" (100 mm)
Material: Polypropylene
Dimensions: 110 x 11 mm
Catalog No. 50-365-013 Supplier Electron Microscopy Sciences Supplier No. 649174
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Boxes are transparent and made from Anti-Electrostatic Polycarbonate.
Use: 4" (100 mm)
Material: Polypropylene
Dimensions: 110 x 11 mm