Learn More
Electron Microscopy Sciences SEM Wafer Holder, 76 mm
SDP
Catalog No. 501928210
Encompass
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 3" (76 mm) diameter wafer.
Size: 76 mm
Catalog No. 50-192-8210 Supplier Electron Microscopy Sciences Supplier No. 7595813
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 3" (76 mm) diameter wafer.
Size: 76 mm