Learn More
Electron Microscopy Sciences SEM Wafer Holder for Notch Style Wafers, 100 mm
SDP
Catalog No. 501928213
Encompass
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length.
Pin: 100 mm
Catalog No. 50-192-8213 Supplier Electron Microscopy Sciences Supplier No. 7595816
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length.
Pin: 100 mm