Learn More
Electron Microscopy Sciences SEM Wafer Holder for Notch Style Wafers, 150 mm
SDP
Catalog No. 501928214
Encompass
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length.
Pin: 150 mm
Catalog No. 50-192-8214 Supplier Electron Microscopy Sciences Supplier No. 7595817
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length.
Pin: 150 mm