Learn More
Electron Microscopy Sciences Wafer-Mount™ 559
SDP
Catalog No. 50997404
Encompass_Preferred
Available in sheet form 10"x10" (254x254 mm). To use, remove the paper backing exposing the adhesive layer. The substrate is positioned on the film and pressed down firmly so that it adheres to the plastic film. No heating is required. The wafer mounted on the plastic film can then be held down using a vacuum manifold or mechanical means for dicing or scribing. After dicing or scribing, the parts can be separated from the wafer-mount 559 film by washing in acetone or MEK. Ideal for holding down silicon wafers, alumina, or glass substrates for scribing.
Catalog No. 50-997-404 Supplier Electron Microscopy Sciences Supplier No. 5040302
Available in sheet form 10"x10" (254x254 mm). To use, remove the paper backing exposing the adhesive layer. The substrate is positioned on the film and pressed down firmly so that it adheres to the plastic film. No heating is required. The wafer mounted on the plastic film can then be held down using a vacuum manifold or mechanical means for dicing or scribing. After dicing or scribing, the parts can be separated from the wafer-mount 559 film by washing in acetone or MEK. Ideal for holding down silicon wafers, alumina, or glass substrates for scribing.