Learn More
Electron Microscopy Sciences Wafer-Mount™ 562
SDP
Supplier:  Electron Microscopy Sciences 5040420
Encompass_Preferred
A dry temporary adhesive film, 0.003" thick, which can be easily cut to size with either a razor or scissor. This mount permits pre-placement of the adhesive film exactly where the user wants it. It melts at 195-210 degree F. After application it can be dissolved rapidly in trichloroethylene or toluene. This material is particularly useful in applications where a pre-formed adhesive film is needed to assure uniform bonding and flatness or in bonding stacks of substrates such as LCD glass slides for slicing. Sheet size 8"x10".
Catalog No. 50-997-407