Learn More
Cleatech WAFER STORAGE DESICCATOR CABNT
SDP
Catalog No. NC2211737
Encompass_Preferred
Wafer Storage Desiccator Cabinet, Clear Acrylic, 38"Wx25"Dx64"H, Eight Chambers, Storage Capacity of 16 Wafer Container (Based on Two boxes per chamber Max. carrier size17"Wx10.5"Dx14.25"H) Stainless Steel Door Frame, with plenum chamber (Not including rack & shelf unless noted)
This item is not returnable. View return policy
Catalog No. NC2211737 Supplier Cleatech Supplier No. 15108O
Add to Cart
Add to Cart
Request a Quote
Wafer Storage Desiccator Cabinet, Clear Acrylic, 38"Wx25"Dx64"H, Eight Chambers, Storage Capacity of 16 Wafer Container (Based on Two boxes per chamber Max. carrier size17"Wx10.5"Dx14.25"H) Stainless Steel Door Frame, with plenum chamber (Not including rack & shelf unless noted)