Learn More
Electron Microscopy Sciences Rubis Wafer Tweezers Style 2LB/4, 128 mm
SDP
Catalog No. 5023939
Encompass_Preferred
Made from SA steel with serrated handles. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers. They have a smooth polished non-glare surface with a satin finish. The tweezers have stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping. No teeth.
OAL: 128 mm
Width: 12.5 mm
Catalog No. 50-239-39 Supplier Electron Microscopy Sciences Supplier No. 7822001
Made from SA steel with serrated handles. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers. They have a smooth polished non-glare surface with a satin finish. The tweezers have stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping. No teeth.
OAL: 128 mm
Width: 12.5 mm