Learn More
Electron Microscopy Sciences Rubis Wafer Tweezers Style 43LB-8 Peek, 138 mm
SDP
Catalog No. 5023932
Encompass_Preferred
For wafers handling. Made of PEEK an ESD safe soft, conductive fiber, with heat resistance up to 250°C. Clean-room compatible.
OAL: 138 mm
Width: 27 mm
Catalog No. 50-239-32 Supplier Electron Microscopy Sciences Supplier No. 7823901
For wafers handling. Made of PEEK an ESD safe soft, conductive fiber, with heat resistance up to 250°C. Clean-room compatible.
OAL: 138 mm
Width: 27 mm