Learn More
Ted Pella Inc Wafer Carrier Tray for 4"/100mm Single Wafer, Natural Polypropylene, 10/pk
SDP
Catalog No. NC0770783
Encompass
These high quality wafer carrier trays are specifically designed for storage, transport and handling of semiconductor wafers, glass wafers or other delicate flat objects. Made from either natural or conductive black polypropylene material and available in sizes from 1" to 6" with maximum grip, minimum contact and minimum pressure. The wafer or disc carriers have a conical base to support the wafers at the edges only. The wafers are held in place and secured by a spider spring and locking cap. Clamshell type wafer carriers are available for 6"/150mm, 8"/200mm and 12"/300mm wafers in clear and conductive black polycarbonate. Internal height is 0.7mm for the 6" and 8" carriers and is 0.8mm for the 12" carriers.
Catalog No. NC0770783 Supplier Ted Pella Inc Supplier No. 139540
These high quality wafer carrier trays are specifically designed for storage, transport and handling of semiconductor wafers, glass wafers or other delicate flat objects. Made from either natural or conductive black polypropylene material and available in sizes from 1" to 6" with maximum grip, minimum contact and minimum pressure. The wafer or disc carriers have a conical base to support the wafers at the edges only. The wafers are held in place and secured by a spider spring and locking cap. Clamshell type wafer carriers are available for 6"/150mm, 8"/200mm and 12"/300mm wafers in clear and conductive black polycarbonate. Internal height is 0.7mm for the 6" and 8" carriers and is 0.8mm for the 12" carriers.