Learn More
Cayman Chemical WAgatoxIn IVAtrIfluoROCt 100ug
SDP
Catalog No. 502987466
Encompass_Preferred
A natural peptide that selectively and irreversibly blocks P-type calcium channels (IC50 <1 nM)
Catalog No. 50-298-7466 Supplier Cayman Chemical Supplier No. 21605100 UG
A natural peptide that selectively and irreversibly blocks P-type calcium channels (IC50 <1 nM)