Learn More
Med Vet International WAGD Outlet, Wall, Chemtron, J0537WAGW, Each
Catalog No. 5000584629
Encompass_Preferred
WAGD (Waste Anesthesia Gas Disposal) Interface. A WAGD outlet is a high volume and pressure suction outlet. In order to be used for gas scavenging for an anesthesia machine, this pressure needs to be stepped down via a WAGD interface. It is mounted on the anesthesia machine pole. 3-way control valve and hoses. Outlets for WAGD WAGD wall Chemtron
Catalog No. 50-005-84629 Supplier Med Vet International Supplier No. JOR537WAGW
WAGD (Waste Anesthesia Gas Disposal) Interface. A WAGD outlet is a high volume and pressure suction outlet. In order to be used for gas scavenging for an anesthesia machine, this pressure needs to be stepped down via a WAGD interface. It is mounted on the anesthesia machine pole. 3-way control valve and hoses. Outlets for WAGD WAGD wall Chemtron