Learn More
Fine Science Tools WAGNER SCISSORS CURVED 12CM
SDP
Catalog No. NC0528921
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 30mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No
Catalog No. NC0528921 Supplier Fine Science Tools Supplier No. 1406912
Add to Cart
Add to Cart
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 30mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No