Learn More
Grainger Little Giant Wagon Truck, Welded Steel, 2,000 lb Capacity, 72 in. x 30 in., 1 ea.
Catalog No. 502784031
Encompass_Preferred
This caster steer wagon is designed for efficient material transport in industrial settings. Constructed with a durable 12-gauge welded steel frame and a smooth solid deck, this unit provides a reliable platform for moving heavy loads up to 2,000 lb. It features a powder-coated finish for resistance to wear and is equipped with a comfortable vinyl-grip T-handle for maneuverability.
  • Load capacity of 2,000 lb
  • Solid deck measuring 72 in. x 30 in.
  • Includes 1-1/2 in. sides for load containment
  • Features two rigid and two swivel casters with 8 in. rubber wheels
  • Durable powder-coated finish on frame and deck
  • Assembled for immediate use
Catalog No. 50-278-4031 Supplier Grainger Supplier No. 19G743
This caster steer wagon is designed for efficient material transport in industrial settings. Constructed with a durable 12-gauge welded steel frame and a smooth solid deck, this unit provides a reliable platform for moving heavy loads up to 2,000 lb. It features a powder-coated finish for resistance to wear and is equipped with a comfortable vinyl-grip T-handle for maneuverability.
  • Load capacity of 2,000 lb
  • Solid deck measuring 72 in. x 30 in.
  • Includes 1-1/2 in. sides for load containment
  • Features two rigid and two swivel casters with 8 in. rubber wheels
  • Durable powder-coated finish on frame and deck
  • Assembled for immediate use