Learn More
MSC Trynex Walk-Behind Sprayer, 12 gal, Electric, Boomless, Powder Coated
Supplier:  MSC 50005107
Encompass
The TurfEx TL80 Walk-Behind Sprayer is an electric-powered, 12-gallon unit for efficient application of liquids like fertilizers, insecticides, herbicides, water, and brine. It features a robust polyethylene tank and a powder-coated metal frame, offering durability and reliable performance for broad area or spot treatments.
  • Electric powered for quiet operation.
  • 12-volt rechargeable battery, sprays up to 120 gallons per charge.
  • Front-mounted, adjustable boomless nozzle (36 to 48 inch spray width).
  • Pistol-grip spray wand with an 8-foot hose for spot treatments.
  • Diaphragm pump: 0.89 GPM at 60 psi.
  • Large pneumatic tires for easy maneuverability.
  • In-line filter prevents clogging.
  • Covers approx. 51,000 sq ft (1.2 acres) per tank.
  • Ideal for liquid fertilizers, insecticides, herbicides, water, and brine.
  • Powder coated metal frame.
Catalog No. 50-283-8003