Learn More
Chemglass Life Sciences Walkabout Insulative Scoop Tray with 0.5L Lab Armor Beads
SDP
Catalog No. 501214792
Encompass_Preferred
Walkabout Insulative Scoop Tray with 0.5L Lab Armor Beads. The new Walkabout Scoop Tray is a quarter of the size of a traditional lab bucket making it easier to use under the hood or in tight bench top spaces. With its superior insulative properties, it keeps your samples and reagents at temperature after removing from the bead bath, Chill Bucket™, or the refrigerator. It lets you feel good about taking your time under the hood. Compatible from -196°C to 93°C.
Catalog No. 50-121-4792 Supplier Chemglass Life Sciences Supplier No. CLS2993001
Walkabout Insulative Scoop Tray with 0.5L Lab Armor Beads. The new Walkabout Scoop Tray is a quarter of the size of a traditional lab bucket making it easier to use under the hood or in tight bench top spaces. With its superior insulative properties, it keeps your samples and reagents at temperature after removing from the bead bath, Chill Bucket™, or the refrigerator. It lets you feel good about taking your time under the hood. Compatible from -196°C to 93°C.