Learn More
Sparkfun Electronics WALL ADAPTER POWER SPLY FOR PI
Catalog No. NC1136985
Encompass_Preferred
Wall Adapter Power Supply - 5.25V DC 2.4A (USB Micro-B). This is a high quality switching ‘wall wart’ AC to DC 5.25V 2400mA USB Micro-B wall power supply manufactured specifically for projects that require a higher level of power. This compact USB AC charger is UL and CSA listed and incorporates a highly regulated output switching power design to assure a constant, accurate DC power delivery.
Catalog No. NC1136985 Supplier Sparkfun Electronics Supplier No. TOL13831
Wall Adapter Power Supply - 5.25V DC 2.4A (USB Micro-B). This is a high quality switching ‘wall wart’ AC to DC 5.25V 2400mA USB Micro-B wall power supply manufactured specifically for projects that require a higher level of power. This compact USB AC charger is UL and CSA listed and incorporates a highly regulated output switching power design to assure a constant, accurate DC power delivery.