Learn More
MSC CAM Spray Pressure Washer, Cold Water, Wall Mount, 3 GPM, 1500 PSI
Catalog No. 502835976
Encompass
This Deluxe Wall Mount Electric Powered Cold Water Pressure Washer has been a popular design for 25 years. It is available with an industrial painted frame or a full stainless steel frame, suitable for tough environments. The cover provides protection for the unit.
  • Dual chemical valve allows selection between two concentrate chemicals and controls proportion.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves, protected by an unloader valve.
  • Low-pressure chemical injection for applying liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.
  • Two hooks for hose and trigger gun storage.
  • Grommets for Q-meg nozzles storage.
Catalog No. 50-283-5976 Supplier MSC Supplier No. 63818850
This Deluxe Wall Mount Electric Powered Cold Water Pressure Washer has been a popular design for 25 years. It is available with an industrial painted frame or a full stainless steel frame, suitable for tough environments. The cover provides protection for the unit.
  • Dual chemical valve allows selection between two concentrate chemicals and controls proportion.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves, protected by an unloader valve.
  • Low-pressure chemical injection for applying liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.
  • Two hooks for hose and trigger gun storage.
  • Grommets for Q-meg nozzles storage.