Learn More
MSC CAM Spray Pressure Washer, Wall Mount, 4 GPM, 2000 PSI
Catalog No. 502837107
Encompass
This electric cold water pressure washer is a popular and robust design, suitable for tough environments with an industrial painted or stainless steel frame. It features easy wall attachment with a mounting bracket and provides storage for hoses, trigger guns, and Q-Meg nozzles.
  • Robust design with industrial painted or stainless steel frame.
  • Protective cover and easy wall attachment with included mounting bracket.
  • Frame includes hooks for hose and trigger gun storage, plus grommets for Q-Meg nozzles.
  • Dual chemical valve for selecting and proportioning two concentrated chemicals.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves.
  • Low-pressure chemical injection for applying liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.
Catalog No. 50-283-7107 Supplier MSC Supplier No. 63820187
Add to Cart
Add to Cart
Request a Quote
This electric cold water pressure washer is a popular and robust design, suitable for tough environments with an industrial painted or stainless steel frame. It features easy wall attachment with a mounting bracket and provides storage for hoses, trigger guns, and Q-Meg nozzles.
  • Robust design with industrial painted or stainless steel frame.
  • Protective cover and easy wall attachment with included mounting bracket.
  • Frame includes hooks for hose and trigger gun storage, plus grommets for Q-Meg nozzles.
  • Dual chemical valve for selecting and proportioning two concentrated chemicals.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves.
  • Low-pressure chemical injection for applying liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.