Learn More
MSC CAM Spray Pressure Washer, Wall Mount, 2 GPM, 1000 psi, Electric, 120 V, 15 A
Catalog No. 502838116
Encompass
This deluxe wall mount cold water electric pressure washer offers 25 years of proven design. It features a choice of industrial painted or full stainless steel frame with a protective cover, designed for easy wall mounting. Integrated hooks provide storage for the hose and trigger gun, while grommets store Q-Meg nozzles. A dual chemical valve enables selection and proportioning of two concentrated chemicals.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves, protected by an unloader valve.
  • Low pressure chemical injection is activated by using the soap tip for concentrated liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.
Catalog No. 50-283-8116 Supplier MSC Supplier No. 63818645
This deluxe wall mount cold water electric pressure washer offers 25 years of proven design. It features a choice of industrial painted or full stainless steel frame with a protective cover, designed for easy wall mounting. Integrated hooks provide storage for the hose and trigger gun, while grommets store Q-Meg nozzles. A dual chemical valve enables selection and proportioning of two concentrated chemicals.
  • Industrial motor is totally enclosed and thermal overload protected.
  • Industrial triplex plunger pump features ceramic plungers and stainless steel valves, protected by an unloader valve.
  • Low pressure chemical injection is activated by using the soap tip for concentrated liquid detergents.
  • Includes 0°, 15°, 25°, and 40° Q-meg tips and a trigger gun.