Learn More
SOMNI Scientific™ Wall Mount Anesthesia System
SDP
Catalog No. 04355316 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
Large Animal
Catalog No. Animal
04-355-316 Large Animal
1 options
Catalog No. 04-355-316 Supplier SOMNI Scientific™ Supplier No. RM17005
Only null left
Add to Cart
Add to Cart
The SOMNI Rebreathing Wallmount Small Animal Anesthesia Machine is made of high-quality materials and has a small footprint. The robust frame provides exceptional stability and the toggle switch conveniently changes from rebreathing to nonrebreathing while circuits stay in place.
  • High quality limited maintenance design
  • Pressure manometer center mounted for immediate visibility
  • Toggle switch: rebreathing to nonrebreathing, circuits stay in place
  • Robust frame provides exceptional stability
  • Popoff valve releases at 2 cm H2O while maintaining passive volume
  • Small footprint, easy assembly
  • Powder coated and anodized frame for easy cleaning
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Animal Large Animal
Type Anesthesia Machine