Learn More
Braintree Scientific Wall Mount CO2 Flow Meter, CO2 (0.5-12 LPM), no bracket
SDP
Catalog No. 501954757
Encompass
Wall Mount CO2 Flow Meter offers precise liters per minute (2-16 LPM) control of CO2 flow. Flow meter ensures minimum gas usage and maximum safety for technicians. Includes 6' hose and quick-disconnect fitting that attaches to Euthanex Lid **Sold for research purposes only and not for human use**
Catalog No. 50-195-4757 Supplier Braintree Scientific Supplier No. EP512A1DS
Wall Mount CO2 Flow Meter offers precise liters per minute (2-16 LPM) control of CO2 flow. Flow meter ensures minimum gas usage and maximum safety for technicians. Includes 6' hose and quick-disconnect fitting that attaches to Euthanex Lid **Sold for research purposes only and not for human use**