Learn More
Andwin Scientific WALL MOUNT PIPETTOR RACK
SDP
Catalog No. 50820168
Encompass_Preferred
Saves space on the bench top. Includes two wood screws for hanging. White acrylic. Holds up to five single channel ErgoOne, Gilson Pipetman, Eppendorf Research plus, Eppendorf Reference, Eppendorf Research, Oxford Benchmate, Labsystems Finnpipette, Sealpette, and Biohit Proline; Rainin Pipet-Lite, Pipet-Plus, and Classic pipettes. Will also hold other brands. Measures 10 1/4" W x 2 3/4" D x 4" H.
Catalog No. 50-820-168 Supplier Andwin Scientific Supplier No. 30960005
Saves space on the bench top. Includes two wood screws for hanging. White acrylic. Holds up to five single channel ErgoOne, Gilson Pipetman, Eppendorf Research plus, Eppendorf Reference, Eppendorf Research, Oxford Benchmate, Labsystems Finnpipette, Sealpette, and Biohit Proline; Rainin Pipet-Lite, Pipet-Plus, and Classic pipettes. Will also hold other brands. Measures 10 1/4" W x 2 3/4" D x 4" H.