Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Wall Mount Splash Guard

Catalog No. 22505227
Change view
Click to view available options
Height (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
22-505-227 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-227 Supplier Marketlab Supplier No. 21648
Add to Cart
Add to Cart

Helps protect floors and counters from splash hazards.

  • Wall-mounted splash guard prevents water from splashing on the floor or counter and minimizes hazards
  • Ideal for stand-alone sinks or sinks with minimal counter space
  • Can also be used as a barrier between workstations
  • Polycarbonate shield with aluminum brackets

Specifications

Lens Coating Uncoated
Height (English) 12 in.
Width (English) 15.87 in.
Product Type Splash Shield
Material Acrylic
Disposable Reusable
Lens Material None
UV Protection None
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.