Learn More
Fisherbrand™ Wall Mounted Cylinder Restraint
Catalog No. 06000497 Shop All Fisherbrand Products
Catalog No. 06-000-497
Add to Cart
Add to Cart
Includes
Nylon Strap and Security Chain
The Fisherbrand™ Wall Mounted Cylinder Restraint can be placed adjacent to eachother to secure a group of cylinders.
  • Cylinder restraint is cast aluminum
  • Has nylon strap and security chain
Specifications
Product Type Cylinder Restraint
Includes Nylon Strap and Security Chain
For Use With (Application) Cylinders
For Use With (Equipment) Cylinders
Device Holds 1 Cylinder
Diameter (Metric) Outer Cylinder Diameter can be 4-14 in.
Height (English) 4.6 in.
Height (Metric) 11.8 cm
Length (English) 8.1 in.
Length (Metric) 20.6 cm