Learn More
Brady™ Wall Mounted Lockout Station: 20 Lock Padlock Board, Spanish
Catalog No. 19103993 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-103-993 Yellow
1 options
Catalog No. 19-103-993 Supplier Brady™ Supplier No. 121506
Add to Cart
Add to Cart
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Wall Mounted Lockout Station is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Color Yellow
Displayed Text ESTACION DE BLOQUEO
Includes Lockout board (board only)
Quantity 1 Ea.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.