Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Wall Shelf
SCI_ED

Catalog No. S37886 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (D x W x H):
12 x 48 x 11.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
S37886 12 x 48 x 11.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37886 Supplier Diversified Spaces™ Supplier No. 250514
Add to Cart
Add to Cart

For storing daily supplies

  • Made from 18 gauge T-304 stainless-steel
  • 12 in. D shelves with 8 in. angle supports
  • 1.5 in. backsplash

Specifications

Product Type Wall and Island Bench
Color Gray
For Use With (Equipment) storing daily supplies
Dimensions (D x W x H) 12 x 48 x 11.5 in.
Material Stainless Steel (18 ga.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.