Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Accessories for accu-jet™ Pipette Controller

Catalog No. 13689552 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
accu-jet S
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-689-552 accu-jet S
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-552 Supplier BRANDTECH™ BRAND™ Supplier No. 26539
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Accessories and replacement parts for use with your BRAND™ accu-jet™ S and pro pipette controller.

For accu-jet™ S:

  • Part No. 26539: Plastic wall support
  • Part No. 26606: Universal AC Adapter, 100-240V / 50-60Hz
  • Part No. 26547: Snape tape

For accu-jet™ pro:

  • Part No. 26540: Plastic wall support

For accu-jet™ S and accu-jet™ S:

  • Part No: 26630: NiMH Battery
  • Part No. 26508: Silicone Adapter, with non-return valve, 54 mm
TRUSTED_SUSTAINABILITY

Specifications

Description Wall mount for accu-jet S
Autoclavable Autoclavable
Color White
Disposable Reusable
For Use With accu-jet S
Material Polypropylene
Number of Channels Variable
Sterility Non-sterile
Type Wall Support
For Use With (Equipment) accu-jet S
Weight (Metric) 21 g
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.