Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Wallerstein Broth (WL) 2mL Ampoules (50 Pack)

Catalog No. 10486144 Shop All GVS Products
Change view
Click to view available options
Broth Type:
Wallerstein Differential Broth
Wallerstein Nutrient Broth
Detects:
Bacterial counts
Yeast growth and counts
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Broth Type Detects
10-486-144 Wallerstein Differential Broth Bacterial counts
10-486-143 Wallerstein Nutrient Broth Yeast growth and counts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-486-144 Supplier GVS™ Supplier No. 10496109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use 2mL ampoule Wallerstein Broth (WL) is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

Description

  • WL Nutrient Broth is for the cultivation and enumeration of yeast and WL Differential Broth is for determination of bacterial count. Use of the medium at pH 5.5 and incubation at 25°C will give reliable counts for brewer’s yeast. Adjustment of the pH to 6.5 and incubation at 30°C allows for the selective growth of baker’s and distiller’s yeast

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
  • 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Use with membrane filters to culture bacteria, yeasts, and molds.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Appearance Blue Nutrient Liquid
Broth Type Wallerstein Differential Broth
Certifications/Compliance Batch-specific quality certificate in each pack
Color Green-blue
Content And Storage 2°C to 8°C
Additive Cyclohexamide
Detects Bacterial counts
Format Ampule
Manufacturing Origin United States
Media Type Wallerstein Differential Broth
Sterility Sterile
Test Type Microbial Cultivation
Fill Volume 2 to 2.4 mL
Formulation Yeast Extract-4.0 gm, Pancreatic Digest of Casein-5.0 gm; Dextrose-50.0 gm,Monopotassium Phosphate-0.55 gm, Potassium Chloride-0.425 gm, Calcium Chloride-0.125 gm, Magnesium Sulfate-0.125 gm,Ferric Chloride- 0.0025 gm, Manganese Sulfate-0.0025 gm, Bromocresol Green-0.022 gm, Cyclohexamide-0.004 gm
For Use With (Application) Isolation and enumeration of bacteria in breweries and industrial fermentation
Includes 50 Ampules, Labeled Foil Bag, Heat Sealer, Batch-specific Quality Certificate
Incubation Conditions plantarum-5°C ±2°C,,S. cerevisiae - 25°C ±2°C
Incubation Time 48 hr. (Organism Dependent)
Material Polypropylene
No. per Pack 50
Packaging Type Ampoules
Quantity 2 mL
Dimensions (Dia. x H) 22 x 57.9 mm
Height (Metric) 57.9 mm
Diameter (Metric) 22 mm
Protein Source Yeast Extract, Casein
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.