Learn More
Strem, An Ascensus Company CAS 894771-28-9. 100mg. (R)-()-1-[(R)-2-(2-Di-35-xylylphosphinophenyl)ferrocenyl]ethyldi-35-xylylphosphine min. 97%. MFCD04117713
SDP
Catalog No. 502110796
Encompass
CAS 894771-28-9. 100mg. (R)-()-1-[(R)-2-(2-Di-35-xylylphosphinophenyl)ferrocenyl]ethyldi-35-xylylphosphine min. 97%. MFCD04117713. Molecular Weight 770.74. Molecular Formula C50H52FeP2. Color/form orange-red solid Strem 26-1555. http//www.strem.com/catalog/v/26-1555/
Catalog No. 50-211-0796 Supplier Strem, An Ascensus Company Supplier No. 261555100MG
CAS 894771-28-9. 100mg. (R)-()-1-[(R)-2-(2-Di-35-xylylphosphinophenyl)ferrocenyl]ethyldi-35-xylylphosphine min. 97%. MFCD04117713. Molecular Weight 770.74. Molecular Formula C50H52FeP2. Color/form orange-red solid Strem 26-1555. http//www.strem.com/catalog/v/26-1555/