Learn More
TARGETMOL CHEMICALS INC WALRYCIN B 10MG
SDP
Catalog No. 502577291
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Walrycin B is a new-type antibacterial compound targeting the WalR response regulator. purity: 99%
Catalog No. 50-257-7291 Supplier TARGETMOL CHEMICALS INC Supplier No. T183210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Walrycin B is a new-type antibacterial compound targeting the WalR response regulator. purity: 99%