Learn More
Medchemexpress LLC Walrycin B | 878419-78-4 | 98.1% | 337.26 | 100 MG
SDP
Catalog No. 5000387702
Encompass_Preferred
Walrycin B is an analogue of toxoflavin and a potent SARS-CoV-2 3CLpro inhibitor. It also acts as a WalR response regulator inhibitor and demonstrates potent activity in inhibiting bacteria growth. This compound is for research use only.
  • Potent SARS-CoV-2 3CLpro inhibitor
  • WalR response regulator inhibitor
  • Exhibits potent activity in inhibiting bacteria growth
  • IC50 of 0.26 μM for SARS-CoV-2 3CLpro
Catalog No. 50-003-87702 Supplier Medchemexpress LLC Supplier No. HY18219100MG
Walrycin B is an analogue of toxoflavin and a potent SARS-CoV-2 3CLpro inhibitor. It also acts as a WalR response regulator inhibitor and demonstrates potent activity in inhibiting bacteria growth. This compound is for research use only.
  • Potent SARS-CoV-2 3CLpro inhibitor
  • WalR response regulator inhibitor
  • Exhibits potent activity in inhibiting bacteria growth
  • IC50 of 0.26 μM for SARS-CoV-2 3CLpro