Learn More
Walter Stern Scalpel Blade Series 300
Catalog No. 17654456 Shop All Walter Stern Inc. Products
Change view
Click to view available options
Size:
# 10
# 11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-654-456 # 10
13-812-52 # 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-654-456 Supplier Walter Stern Supplier No. 320001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits handles No.3, 3L, 4, 4L and 7
  • Made of Stainless-steel
  • Sterile and Individual sealed
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
Size # 10
For Use With (Equipment) Fits handles # 3, # 3L, # 4, # 4L and # 7
Sterility Sterile