Learn More
Walter Stern Series 300 Scalpel Handles
Catalog No. 1381254 Shop All Walter Stern Inc. Products
Change view
Click to view available options
Size:
# 3
Catalog No. Size
13-812-54 # 3
1 options
This item is not returnable. View return policy
Catalog No. 13-812-54 Supplier Walter Stern Supplier No. 320051
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Blade Material Stainless Steel
Size # 3
For Use With (Equipment) Fits Blades # 10, # 11, # 12 and # 15