Learn More
Walter Stern Vernier Caliper Nickel-Plated Dual Scale Caliper
Catalog No. 121251 Shop All Walter Stern Inc. Products
Change view
Click to view available options
Material:
Plated Nickel
Catalog No. Material
12-125-1 Plated Nickel
1 options
Catalog No. 12-125-1 Supplier Walter Stern Supplier No. 602B
Add to Cart
Add to Cart
Rustproof and measures both inside and outside dimensions
Caliper has a 12cm or 5 in. scale with thumb roller adjustment and lever lock. Two scales total: one reads to 1/128 in. and the other reads to 1/10mm.
Specifications
Material Plated Nickel
For Use With (Application) Measures both inside and outside dimensions
Includes Thumb roller adjustment and lever lock, two scales
Product Type Dual Scale Vernier Caliper