Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alere Clearview™ hCG Rapid Lateral Flow Test Kits

Catalog No. WA6028KCV Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Clearview Hcg Controls
Clearview Hcg Ii Urine Pk/50
Clrview Hcg Combo Ref Ln 30/pk
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
WA6028KCV Clrview Hcg Combo Ref Ln 30/pk
WA6007KCV Clearview Hcg Ii Urine Pk/50
WA-6009KCV Clearview Hcg Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. WA6028KCV Supplier Alere Supplier No. 6028KCV

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Results in five minutes or less

•    CLIA-waived for urine; non-waived for serum
•    Simple, one-step procedure
•    Tests for urine or serum
•    Results in five minutes or less
•    Built-in controls
•    Sensitivity: 20mIU/mL for urine; 10mIU/mL for serum
•    Specificity: 100%
•    Room temperature storage with up to 24 months dating

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.