Learn More
Apexbio Technology LLC Wang Resin 201058-08-4 100g
SDP
Catalog No. 5000093162
Encompass_Preferred
Wang Resin is a polymeric solid-support matrix used as a substrate for Fmoc-based solid-phase peptide synthesis The resin consists of cross-linked polystyrene particles functionalized with p-hydroxybenzyl alcohol groups enabling covalent anchoring of amino acids through ester bond formation Sequential coupling and deprotection cycles facilitate stepwise peptide chain extension After synthesis peptides are cleaved from Wang Resin under mild acidic conditions producing free peptides for further analysis Wang Resin is widely employed in peptide-based research applications including peptide discovery structural analysis receptor binding assays and bioactive peptide development
Catalog No. 50-000-93162 Supplier Apexbio Technology LLC Supplier No. A6776100000
Wang Resin is a polymeric solid-support matrix used as a substrate for Fmoc-based solid-phase peptide synthesis The resin consists of cross-linked polystyrene particles functionalized with p-hydroxybenzyl alcohol groups enabling covalent anchoring of amino acids through ester bond formation Sequential coupling and deprotection cycles facilitate stepwise peptide chain extension After synthesis peptides are cleaved from Wang Resin under mild acidic conditions producing free peptides for further analysis Wang Resin is widely employed in peptide-based research applications including peptide discovery structural analysis receptor binding assays and bioactive peptide development