Learn More
Apexbio Technology LLC Wang Resin 201058-08-4 25g
SDP
Catalog No. 5000093610
Encompass_Preferred
Wang Resin is a polymeric solid-support matrix used as a substrate for Fmoc-based solid-phase peptide synthesis The resin consists of cross-linked polystyrene particles functionalized with p-hydroxybenzyl alcohol groups enabling covalent anchoring of amino acids through ester bond formation Sequential coupling and deprotection cycles facilitate stepwise peptide chain extension After synthesis peptides are cleaved from Wang Resin under mild acidic conditions producing free peptides for further analysis Wang Resin is widely employed in peptide-based research applications including peptide discovery structural analysis receptor binding assays and bioactive peptide development
Catalog No. 50-000-93610 Supplier Apexbio Technology LLC Supplier No. A677625000
Add to Cart
Add to Cart
Wang Resin is a polymeric solid-support matrix used as a substrate for Fmoc-based solid-phase peptide synthesis The resin consists of cross-linked polystyrene particles functionalized with p-hydroxybenzyl alcohol groups enabling covalent anchoring of amino acids through ester bond formation Sequential coupling and deprotection cycles facilitate stepwise peptide chain extension After synthesis peptides are cleaved from Wang Resin under mild acidic conditions producing free peptides for further analysis Wang Resin is widely employed in peptide-based research applications including peptide discovery structural analysis receptor binding assays and bioactive peptide development