Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ War of the Acids Chemistry Demonstration Kit
GREENER_CHOICESCI_ED

Catalog No. S20638 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S20638 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S20638 Supplier Innovating Science™ Supplier No. IS7044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Designed to visualize the differences between strong and weak acids

Activities include demonstrations of differ­ences in pH values, effect of conjugate base and vigor of reactions.

This kit contains an easy to follow teachers manual and enough materials to perform 7 demonstrations.

Kit includes:
  • 650mL Hydrochloric Acid
  • 450mL Acetic Acid
  • 30mL Low Range pH Indicator
  • 115g Sodium Acetate
  • 85g Sodium Chloride
  • 290g Calcium Carbonate
  • 7 Plastic Pipettes
  • Color Code Indicator Chart

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Demonstrating of differ­ences in pH values, effect of conjugate base and vigor of reactions
Includes
  • 650mL Hydrochloric Acid
  • 450mL Acetic Acid
  • 30mL Low Range pH Indicator
  • 115g Sodium Acetate
  • 85g Sodium Chloride
  • 290g Calcium Carbonate
  • 7 Plastic Pipettes
  • Color Code Indicator Chart
Quantity 1
Class Size 7 Demonstrations
Product Type Chemistry demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.