Learn More
American Radiolabeled Chemicals Inc WARFARIN 3H 250UC
SDP
Catalog No. 50901197
Encompass_Preferred
Warfarin-ring-3H 250uCi in ethanol, 1 mCi/ml, S.A. 20-40 Ci/mmol, M.W. 308.33, Shipped in Dry Ice
Catalog No. 50-901-197 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1765250UC
Warfarin-ring-3H 250uCi in ethanol, 1 mCi/ml, S.A. 20-40 Ci/mmol, M.W. 308.33, Shipped in Dry Ice