Learn More
Cayman Chemical -WarfarIn-d5 500ug
SDP
Catalog No. 502936223
Encompass_Preferred
An internal standard for the quantification of warfarin by GC- or LC-MS
Catalog No. 50-293-6223 Supplier Cayman Chemical Supplier No. 25432500 UG
An internal standard for the quantification of warfarin by GC- or LC-MS