Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Warfarin Sodium (contains Isopropyl Alcohol) 98.0+%, TCI America™
SDP

Catalog No. W00055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
W00055G 5 g
W000525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. W00055G Supplier TCI America Supplier No. W00055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129-06-6
Molecular Formula C19H18NaO4
Molecular Weight (g/mol) 333.339
MDL Number MFCD00083223
InChI Key MBNHFFMQYKZCLK-UHFFFAOYSA-N
Synonym 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt
PubChem CID 131664125
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one;molecular hydrogen;sodium
SMILES [HH].CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O.[Na]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1544
Formula Weight 330.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Warfarin Sodium (contains Isopropyl Alcohol)
RTECSNumber GN4725000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.