Learn More
Cayman Chemical Warfarn 10mg
SDP
Catalog No. 502955179
Encompass_Preferred
Antgen purfed eggshell protens (Zrp and small amounts of zona pellucda protens) from Atlantc salmon (S salar) Host mouse clone MN2B4 CrossReactvty ( ) Atlantc salmon (S salar) (reacts prmarly wth B monomer) ranbow trout (O mykss) and Arctc char (S alpnus) Zrp Applcaton(s) WB and ELSA sotype gG1K
Catalog No. 50-295-5179 Supplier Cayman Chemical Supplier No. 13526 10MG
Antgen purfed eggshell protens (Zrp and small amounts of zona pellucda protens) from Atlantc salmon (S salar) Host mouse clone MN2B4 CrossReactvty ( ) Atlantc salmon (S salar) (reacts prmarly wth B monomer) ranbow trout (O mykss) and Arctc char (S alpnus) Zrp Applcaton(s) WB and ELSA sotype gG1K