Learn More
Description
Includes
Base, container, 3-wire grounded cord and plug
Commercial-grade units for demanding mixing and blending applications
- Cloverleaf design
- Choice of glass and/or stainless-steel containers
- Sturdy, diecast metal bases and stainless-steel blades
- All durably constructed for years of dependable service
- Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
- Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
- Two-piece vinyl and styrene lids on all models
Order Info
Electrical requirements for all models: 115/120V 50/60Hz
Certifications
UL listed (Select models only; see ordering chart)
Specifications
Specifications
| Amperage | 3.6 A |
| Speed | 18,000, 22,000 rpm |
| Voltage | 115/120 V |
| Electrical Requirements | 115/120 V, 50/60 Hz |
| Horsepower | 0.4 hp |
| Capacity (Metric) | 940 mL |
| Sample Volume (English) | 32 oz. |
| Depth (English) | 7 in. |
| Depth (Metric) | 17.78 cm |
| Dimensions (L x W x H) | 9.75 x 8 x 16 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.