Learn More
New England Biolabs, Inc. WarmStart LAMP Kit (DNA & RNA) – 500 reactions
SDP
Supplier:  New England Biolabs, Inc. E1700L
Encompass
The WarmStart LAMP Kit (DNA & RNA) is designed to provide a simple, one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provides rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase. This kit is supplied with the WarmStart LAMP 2X Master Mix, which contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized LAMP buffer solution. Both Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase have been engineered for improved performance in LAMP and RT-LAMP reactions. A fluorescent dye is also supplied to enable real-time fluorescence measurement of LAMP. The WarmStart LAMP Kit is compatible with multiple detection methods, including turbidity detection, real-time fluorescence detection, and end-point visualization.
Catalog No. NC1345107