Learn More
Med Vet International Slide Warmer, with Hinged Cover, Large
Catalog No. 503046620
Encompass_Preferred
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display.
Catalog No. 50-304-6620 Supplier Med Vet International Supplier No. XH2004
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display.