Learn More
Med Vet International Slide Warmer, Small with Hinged Cover
Catalog No. 503046441
Encompass_Preferred
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display.
Catalog No. 50-304-6441 Supplier Med Vet International Supplier No. XH2005
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display.