Learn More
Med Vet International Warming Air Bag, Small, 30" x 20", J1034
Catalog No. 5000583737
Encompass_Preferred
Keeping patients warm during surgery and recovery has always been a challenge. Electric pads have a risk of overheating, circulating water blankets are easily damaged. Air blankets are a safe, economical solution to this problem. They are made of a silky polyester fabric with an inlay of nylon waterproof material to keep the airflow circulating upward towards the patient. These blankets are machine washable (hang to dry) therefore, much more economical than other air blankets on the market. There is a 5" diameter round opening with a nylon cord to secure the blanket to any of the major brands of warm air convection units.
Catalog No. 50-005-83737 Supplier Med Vet International Supplier No. JOR1034
Add to Cart
Add to Cart
Keeping patients warm during surgery and recovery has always been a challenge. Electric pads have a risk of overheating, circulating water blankets are easily damaged. Air blankets are a safe, economical solution to this problem. They are made of a silky polyester fabric with an inlay of nylon waterproof material to keep the airflow circulating upward towards the patient. These blankets are machine washable (hang to dry) therefore, much more economical than other air blankets on the market. There is a 5" diameter round opening with a nylon cord to secure the blanket to any of the major brands of warm air convection units.