Learn More
Braintree Scientific Warming Blanket, 8"""" x 6""""
SDP
Catalog No. 501956102
Encompass
Warming Blanket Flexible blankets for use with the TCAT-2LV and TCAT-2DF controllers. These blankets use far infrared radiation to penetrate deep into the body. The smaller blanket 8 x 6 is suitable for mice and small rats. The larger blanket 14 x 14 is suitable for adult rats and guinea pigs. Product Specifications: Model no. WB-1 WB-2 Dimensions 8" x 6" 14" x 14" Power 9.5 V @ 1.5 amps 9.5 V @ 1.5 amps Over Temperature Protection 100 Ohm RTD 100 Ohm RTD Lead Length 3 ft. 3 ft. Material Antibacterial Washable Surface Antibacterial Washable Surface **Sold for research purposes only and not for human use**
Catalog No. 50-195-6102 Supplier Braintree Scientific Supplier No. WB1
Warming Blanket Flexible blankets for use with the TCAT-2LV and TCAT-2DF controllers. These blankets use far infrared radiation to penetrate deep into the body. The smaller blanket 8 x 6 is suitable for mice and small rats. The larger blanket 14 x 14 is suitable for adult rats and guinea pigs. Product Specifications: Model no. WB-1 WB-2 Dimensions 8" x 6" 14" x 14" Power 9.5 V @ 1.5 amps 9.5 V @ 1.5 amps Over Temperature Protection 100 Ohm RTD 100 Ohm RTD Lead Length 3 ft. 3 ft. Material Antibacterial Washable Surface Antibacterial Washable Surface **Sold for research purposes only and not for human use**