Learn More
Shockwatch Div Mri WarmMarkAscending Time-Temperature Indicator 5C/41F
SDP
Catalog No. NC2202792
Encompass_Preferred
WarmMark time-temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably high temperature levels, allowing you to determine if product quality has been compromised. Monitor time above the device threshold and indicate if temperature exceeded for the specified time. Indicators are field-armable by pulling a tab. Temperature threshold: 5C/41F. Accuracy +/-1C. Above temperature exposure time: 8 hours. Shelf life 2 years. Single-use.
This item is not returnable. View return policy
Catalog No. NC2202792 Supplier Shockwatch Div Mri Supplier No. WM 5/41
WarmMark time-temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably high temperature levels, allowing you to determine if product quality has been compromised. Monitor time above the device threshold and indicate if temperature exceeded for the specified time. Indicators are field-armable by pulling a tab. Temperature threshold: 5C/41F. Accuracy +/-1C. Above temperature exposure time: 8 hours. Shelf life 2 years. Single-use.